missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534220
This item is not returnable.
View return policy
Description
SSTR2 Polyclonal Antibody for IHC (P)
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
| SSTR2 | |
| Polyclonal | |
| Unconjugated | |
| SSTR2 | |
| Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100052744, 100753171, 20606, 282083, 395305, 403472, 497059, 54305, 6752 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P30680, P30874, P30875, P34993, P34994, Q49LX6 | |
| SSTR2 | |
| Synthetic 16 amino acid peptide from C-terminus of human SSTR2. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Frog, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction