missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA316736
This item is not returnable.
View return policy
Description
SSTR2 Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, Flow, IP, IM
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
| SSTR2 | |
| Polyclonal | |
| Unconjugated | |
| SSTR2 | |
| Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P30680, P30874, P30875, P34994 | |
| SSTR2 | |
| A synthetic peptide made to the C-terminus of rat Somatostatin Receptor 2. | |
| RUO | |
| 20606, 497059, 54305, 6752 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction