missing translation for 'onlineSavingsMsg'
Learn More

SSTK-IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-33923

 View more versions of this product

Product Code. 18488941

  • £571.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SSTK-IP Polyclonal antibody specifically detects SSTK-IP in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SSTK-IP
Polyclonal
Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Q96A04
Rabbit
Immunogen affinity purified
RUO
128229
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry
Unconjugated
PBS (pH 7.2) and 40% Glycerol
C1orf182, chromosome 1 open reading frame 182, MGC26877, RP11-443G18.5, SIP, SSTK-interacting protein (SSTK-IP), SSTK-IP
This antibody was developed against a recombinant protein corresponding to amino acids: MERHTSHPNRKVPAKEEANAVPLCRAKPSPSYINLQASSPPATFLNIQTTKLPSVDHKPKECLG
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.