missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer SST™ II Advance Tubes
Contain an inert gel barrier that separates the serum from the blood clot during centrifugation, preventing contamination of the serum.
Brand: BD Vacutainer 368965
Additional Details : Weight : 0.30000kg
Description
- Contain an inert gel barrier that separates the serum from the blood clot during centrifugation, preventing contamination of the serum.
- Stable barrier between serum and clotted blood during transportation and storage, leading to better analyte stability.
- Routine analytes in clinical chemistry such as potassium are still stable after a week of storage at 2-8°C.
- Clinical evaluation of special chemistries such as therapeutic drugs, proteins, peptides, steroids and vitamins demonstrates a high degree of stability.
- Enable faster centrifugation times of 5 minutes at 3000 g.
- Better sample quality.
- Optimization of the work flow: short centrifugation time, sample processing and archiving in the primary tube.
- No requirement for secondary tubes, reducing the possibility of misidentification.
- Minimum recommended clotting time is 30 minutes.
- Serum separation tubes should be gently inverted 180° and back 5-6 times.
Specifications
Silica (Clot Activator), Gel Separator | |
Hemogard | |
Red | |
Venous blood sampling, serum analysis |
PET | |
3.5 mL | |
13 x 75 mm | |
Paper |