missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSSCA1 Antibody (OTI2F5), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74359PECY7
This item is not returnable.
View return policy
Description
SSSCA1 Monoclonal specifically detects SSSCA1 in Human samples. It is validated for Flow Cytometry.
Specifications
| SSSCA1 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ZNRD2 | |
| Full length recombinant protein of human SSSCA1 (NP_006387) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI2F5 | |
| Flow Cytometry | |
| Autoantigen p27, centromeric autoantigen (27kD), p27, Sjoegren syndrome/scleroderma autoantigen 1, Sjogren syndrome/scleroderma autoantigen 1, Sjogren′s syndrome/scleroderma autoantigen 1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 10534.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction