missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSSCA1 Antibody (OTI2F5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74359CL1
This item is not returnable.
View return policy
Description
SSSCA1 Monoclonal antibody specifically detects SSSCA1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| SSSCA1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Autoantigen p27, centromeric autoantigen (27kD), p27, Sjoegren syndrome/scleroderma autoantigen 1, Sjogren syndrome/scleroderma autoantigen 1, Sjogren's syndrome/scleroderma autoantigen 1 | |
| Full length human recombinant protein of human SSSCA1 (NP_006387) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10534 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI2F5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction