missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSRP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18676-50UG
This item is not returnable.
View return policy
Description
SSRP1 Polyclonal antibody specifically detects SSRP1 in Human samples. It is validated for Western Blot
Specifications
| SSRP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Chromatin-specific transcription elongation factor 80 kDa subunit, cisplatin-DNA SSRP, facilitates chromatin remodeling 80 kDa subunit, Facilitates chromatin transcription complex 80 kDa subunit, Facilitates chromatin transcription complex subunit SSRP1, FACT, FACT complex subunit SSRP1, FACT80FACT 80 kDa subunit, FACTp80, high mobility group box, hSSRP1, Recombination signal sequence recognition protein 1, structure specific recognition protein 1, Structure-specific recognition protein 1, T160 | |
| Polyclonal antibody raised in rabbit against human SSRP1 (Structure-specific recognition protein 1), the GST tagged N-terminal part of the protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 2 mg/ml | |
| Western Blot 1:1000 | |
| Q08945 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 6749 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction