missing translation for 'onlineSavingsMsg'
Learn More

SSR3 Rabbit, anti-Human, Polyclonal, Proteintech

Product Code. 18097275 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18097275 20 μL 20µL
18050596 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18097275 Supplier Proteintech Supplier No. 308511AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The SSR3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SSR3. This antibody recognizes human, mouse, rat antigen. The SSR3 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen SSR3
Applications Western Blot, ELISA
Concentration 300 μg/mL
Gene Symbols SSR gamma, SSR3, TRAP gamma, TRAPG
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 6747
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.