Learn More
SSR3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30851-1-AP-150UL
This item is not returnable.
View return policy
Description
The SSR3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SSR3. This antibody recognizes human, mouse, rat antigen. The SSR3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SSR3 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| SSR gamma, SSR3, TRAP gamma, TRAPG | |
| 150 μL | |
| 6747 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |