missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAP alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552811
This item is not returnable.
View return policy
Description
Immunogen sequence: ESRKRKRPIQ KVEMGTSSQN DVDMSWIPQE TLNQINKASP RRLPRKRAQK RSVGSDE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.
Specifications
| TRAP alpha | |
| Polyclonal | |
| Unconjugated | |
| SSR1 | |
| 2510001K09Rik; 6330400D04; Ac2-238; AI159733; AI452176; cb758; DKFZp781N23103; liver regeneration-related protein LRRG137; PGP35; PSEC0262; signal sequence receptor subunit 1; signal sequence receptor subunit alpha; signal sequence receptor, alpha; signal sequence receptor, alpha (translocon-associated protein alpha); SSR; SSR alpha subunit; SSR1; SSRA precursor protein; SSR-alpha; translocon associated protein alpha; translocon-associated protein alpha; translocon-associated protein alpha subunit; translocon-associated protein subunit alpha; TRAP alpha; TRAPA; TRAP-alpha; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107513, 361233, 6745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43307, Q7TPJ0, Q9CY50 | |
| SSR1 | |
| Recombinant protein corresponding to Human SSR1. Recombinant protein control fragment (Product #RP-90181). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction