missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSPN, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00008082-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MGKNKQPRGQQRQGGPPAADAAGPDDMEPKKGTGAPKECGEEEPRTCCGCRFPLLLALLQLALGIAVTVVGFLMASISSSLLVRDTPFWAGIIVCLVAYLGLFMLCVSYQVDERTCIQFSMKLLYFLLSALGLTVCVLAVAFAAHHYSQLTQFTCETTLDSCQCKLPSSEPLSRTFVYRDVTDCTSVTGTFKLFLLIQMILNLVCGLVCLLACFVMWKHRYQVFYVGVRICSLTASEGPQQKISpecifications
| SSPN | |
| Polyclonal | |
| No additive | |
| NM_005086.3 | |
| SSPN | |
| SSPN (NP_005077.2, 1 a.a. ∼ 243 a.a) full-length human protein. | |
| RUO | |
| 8082 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| sarcospan (Kras oncogene-associated gene) | |
| DAGA5/KRAG/NSPN/SPN1/SPN2/nanospan | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction