missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566842
This item is not returnable.
View return policy
Description
Immunogen sequence: KHRYQVFYVGV RICSLTASEG PQQKI Highest antigen sequence identity to the following orthologs - mouse 65%, rat 73%.
Component of the dystrophin-glycoprotein complex (DGC), a complex that spans the muscle plasma membrane and forms a link between the F-actin cytoskeleton and the extracellular matrix. Preferentially associates with the sarcoglycan subcomplex of the DGC.
Specifications
| SSPN | |
| Polyclonal | |
| Unconjugated | |
| SSPN | |
| DAGA5; kirsten-ras-associated protein; KRAG; Kras oncogene-associated; K-ras oncogene-associated protein; microspan; nanospan; NSPN; RGD1559723; sarcospan; sarcospan (Kras oncogene-associated gene); SPN1; SPN2; SSPN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14714 | |
| SSPN | |
| Recombinant Human SSPN. Recombinant protein control fragment (Product #RP-107496). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction