missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssPA/Stringent Starvation Protein A Rabbit anti-E. coli, Clone: 113, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | ssPA/Stringent Starvation Protein A |
|---|---|
| Clone | 113 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254078
|
Novus Biologicals
NBP2-90173-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18249057
|
Novus Biologicals
NBP2-90173-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ssPA/Stringent Starvation Protein A Monoclonal specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA.Specifications
| ssPA/Stringent Starvation Protein A | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 944744 | |
| IgG | |
| Protein A purified |
| 113 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| E. coli | |
| ECK3218, Pog, ssp, stringent starvation protein A | |
| Recombinant E. coli ssPA/Stringent Starvation Protein A | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title