missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssPA/Stringent Starvation Protein A Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06079AF405
This item is not returnable.
View return policy
Description
ssPA/Stringent Starvation Protein A Polyclonal specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA.
Specifications
| ssPA/Stringent Starvation Protein A | |
| Polyclonal | |
| ELISA | |
| ECK3218, Pog, ssp, stringent starvation protein A | |
| Produced in rabbits immunized with purified, recombinant Escherichia coli Escherichia coli ssPA/Stringent Starvation Protein A (P0ACA3; Met 1-Ser 212). | |
| 0.1 mL | |
| Primary | |
| E. coli | |
| Purified |
| ELISA | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 944744 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction