missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssPA/Stringent Starvation Protein A Antibody (113), Alexa Fluor™ 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
ssPA/Stringent Starvation Protein A Monoclonal specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | ssPA/Stringent Starvation Protein A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 113 |
| Conjugate | Alexa Fluor 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ECK3218, Pog, ssp, stringent starvation protein A |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant E. coli ssPA/Stringent Starvation Protein A (P0ACA3; Met1-Ser212). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?