missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ sso7d Polyclonal Antibody
GREENER_CHOICE

Product Code. 17958712
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958712 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17958712 Supplier Invitrogen™ Supplier No. PA5144493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

sso7d Polyclonal Antibody for Western Blot, ELISA

Can constrain negative DNA supercoils. May be involved in maintaining the integrity of the genome at high temperature (By similarity). Stimulates the Holliday junction cleavage activity of Hjc (PubMed:11709558).
TRUSTED_SUSTAINABILITY

Specifications

Antigen sso7d
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene sso7d
Gene Accession No. P39476
Gene Alias sso7d-1
Gene Symbols sso7d
Host Species Rabbit
Immunogen Recombinant Sulfolobus solfataricus DNA-binding protein 7d (sso7d) (2-64AA).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72911125
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.