Learn More
Invitrogen™ SSNA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589201
Description
Immunogen sequence: MTQQGAALQN YNNELVKCIE ELCQKREELC RQIQEEEDEK QRLQNEVRQL TEKLARVNEN LARKIASRNE FDRTIAETEA AYLKILESSQ TLLSVLKREA GNLTKATAPD QKSSGGRDS; Positive Samples: Mouse spleen; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.
SSNA1 (Sjoegren syndrome nuclear autoantigen 1), also known as N14 or NA-14 (nuclear autoantigen of 14 kDa), is a 119 amino acid nuclear and cytoplasmic protein that is widely expressed and belongs to the SSNA1 family. While most highly expressed in testis, SSNA1 is found at lower levels in peripheral blood leukocytes, spleen, colon, thymus, ovary, prostate and small intestine, and is thought to be associated with microtubule structures. SSNA1 contains an N-terminal dimeric coiled-coil domain, a C-terminal basic domain and may target spastin activity at the centrosome. The gene encoding SSNA1 maps to human chromosome 9, which houses over 900 genes and comprises nearly 4% of the human genome.
Specifications
| SSNA1 | |
| Polyclonal | |
| Unconjugated | |
| Ssna1 | |
| 1110003H09Rik; 1190004J23Rik; AA934184; MNCb-2711; N14; NA14; NA-14; Nuclear autoantigen of 14 kDa; Sjoegren syndrome nuclear autoantigen 1; Sjoegren syndrome nuclear autoantigen 1 homolog; Sjogren syndrome nuclear autoantigen 1; Sjogren's syndrome nuclear autoantigen 1; SS nuclear autoantigen 1; Ssna1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 311802, 68475, 8636 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43805, Q9JJ94 | |
| Ssna1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-119 of human SSNA1 (NP_0037222). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.