missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SSNA1 Polyclonal antibody specifically detects SSNA1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | SSNA1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | NA-14, nuclear autoantigen of 14 kDa, Sjogren syndrome nuclear autoantigen 1 |
| Host Species | Rabbit |
| Immunogen | Sjogren syndrome nuclear autoantigen 1. (Uniprot# O43805) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?