missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101788
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SSH3.
The ADF (actin-depolymerizing factor)/cofilin family is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo.
Specifications
| SSH3 | |
| Polyclonal | |
| Unconjugated | |
| Ssh3 | |
| BC028922; hSSH-3L; mSSH-3L; Protein phosphatase Slingshot homolog 3; slingshot 3; slingshot homolog 3; slingshot homolog 3 (Drosophila); slingshot protein phosphatase 3; slingshot-3L; slingshot-like 3; Ssh3; SSH-3; Ssh3l; SSH-3L; SSH-like protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 245857, 365396, 54961 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XIS1, Q8K330, Q8TE77 | |
| Ssh3 | |
| A synthesized peptide derived from human SSH3(Accession Q8TE77), corresponding to amino acid residues T356-G406. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction