missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSH3 Antibody (6F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00054961-M01
This item is not returnable.
View return policy
Description
SSH3 Monoclonal antibody specifically detects SSH3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
Specifications
| SSH3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 3.1.3.16, EC 3.1.3.48, FLJ10928, FLJ20515, FLJ42240, hSSH-3L, protein phosphatase Slingshot homolog 3, slingshot 3, slingshot homolog 3 (Drosophila), SSH3, SSH3L, SSH-3L, SSH-like protein 3 | |
| SSH3 (NP_060327, 293 a.a. ~ 391 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SKEIRQALELRLGLPLQQYRDFIDNQMLLLVAQRDRASRIFPHLYLGSEWNAANLEELQRNRVTHILNMAREIDNFYPERFTYHNVRLWDEESAQLLPH | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 6F9 | |
| Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Sandwich ELISA | |
| NP_060327 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 54961 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction