missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSH1 Antibody (1A5C8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52569-0.025mg
This item is not returnable.
View return policy
Description
SSH1 Monoclonal specifically detects SSH1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| SSH1 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:10000, CyTOF-ready | |
| EC 3.1.3.16, EC 3.1.3.48, FLJ21928, hSSH-1L, KIAA1298FLJ38102, protein phosphatase Slingshot homolog 1, slingshot 1, slingshot homolog 1 (Drosophila), SSH1L, SSH-1L, SSH-like protein 1 | |
| Mouse | |
| 115.5 kDa | |
| 0.025 mg | |
| Phospho Specific | |
| 54434 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 1A5C8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SSH1 | |
| Synthesized peptide fragment of human SLINGSHOT-1L (AA: 1032-1044 ) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction