Learn More
SSEA5 Monoclonal Antibody (8E11-SSEA5), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£64.25 - £182.00
Specifications
Antigen | SSEA5 |
---|---|
Clone | 8E11-SSEA5 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15218107
|
Affymetrix eBioscience
14-8857-82 |
100 μg |
£182.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15208107
|
Affymetrix eBioscience
14-8857-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.Specifications
SSEA5 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Primary | |
Antibody |
8E11-SSEA5 | |
Unconjugated | |
Mouse | |
Human | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.