missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSEA4 Monoclonal Antibody (MC-813-70)
Description
SSEA4 Monoclonal Antibody for Flow, ICC/IF
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications
Specifications
| Antigen | SSEA4 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MC-813-70 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide |
| Gene Alias | SSEA-4; stage-specific embryonic antigen 4 |
| Host Species | Mouse |
| Immunogen | human embryonal carcinoma cell line 2102Ep. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?