missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSEA1 Monoclonal Antibody (MC-480)

Product Code. 12807752
Change view
Click to view available options
Quantity:
100 μg
20 μL
Unit Size:
100µg
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807752 20 μL 20µL
12887742 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12807752 Supplier Invitrogen™ Supplier No. MA1022X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSEA1 Monoclonal Antibody for Flow, ICC/IF, IHC (F), IHC (P)

CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone MC-480
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene FUT4
Gene Accession No. P22083, Q11127
Gene Alias FUTIV; Lewis X; SSEA-1
Gene Symbols FUT4
Host Species Mouse
Immunogen BALB/c mouse immunized with F9 teratocarcinoma stem cells (X-irradiated).
Purification Method Affinity chromatography - MBP
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14345, 2526
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.