missing translation for 'onlineSavingsMsg'
Learn More

SSEA-5 Mouse, anti-Human, PE, Clone: 8e11, STEMCELL Technologies

Product Code. 17118801
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17118801 25 Tests 25 tests
17108801 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17118801 Supplier Stemcell Technologies Supplier No. 60063PE.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
  • The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures

Specifications

Antigen SSEA-5
Applications Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 8e11
Conjugate PE
Host Species Mouse
Quantity 25 Tests
Primary or Secondary Primary
Target Species Human
Isotype IgG1 κ
Cell Type Pluripotent Stem Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.