Learn More
Invitrogen™ SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£169.00 - £443.00
Specifications
| Antigen | SSEA4 |
|---|---|
| Clone | eBioMC-813-70 (MC-813-70) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15520057
|
Invitrogen™
50-8843-80 |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15530057
![]() |
Invitrogen™
50-8843-82 |
100 μg |
£443.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stage-specific embryonic antigen 4 (SSEA-4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.Specifications
| SSEA4 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgG3 | |
| Affinity chromatography | |
| Antibody |
| eBioMC-813-70 (MC-813-70) | |
| Flow Cytometry, Immunocytochemistry | |
| eFluor 660 | |
| Mouse | |
| Human, Mouse, Canine, Rabbit, Monkey, Chicken | |
| SSEA-4; stage-specific embryonic antigen 4 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.