missing translation for 'onlineSavingsMsg'
Learn More

SSEA-4 Antibody (MC-813-70), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18427316 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427316 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427316 Supplier Novus Biologicals Supplier No. NBP226644MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSEA-4 Monoclonal antibody specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Flow Cytometry, Flow (Cell Surface)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-4
Applications Flow Cytometry
Classification Monoclonal
Clone MC-813-70
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation 50mM Sodium Borate
Gene Alias SSEA-4;stage-specific embryonic antigen 4
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Canine, Chicken, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.