missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-4 Antibody (MC-813-70) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1435JF669
This item is not returnable.
View return policy
Description
SSEA-4 Monoclonal antibody specifically detects SSEA-4 in Human,Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| SSEA-4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| SSEA-4;stage-specific embryonic antigen 4 | |
| 2120Ep human embryonal carcinoma cell line | |
| RUO | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| MC-813-70 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction