missing translation for 'onlineSavingsMsg'
Learn More

SSEA-3 Rat, anti-Mouse, PE, Clone: MC-631, STEMCELL Technologies

Product Code. 17178781
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17178781 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17178781 Supplier Stemcell Technologies Supplier No. 60061PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

  • The MC-631 antibody reacts with a galactosylgloboside epitope on the stage-specific embryonic antigen-3 (SSEA-3), which is expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey ES cell lines
  • No immunoreactivity is evident with undifferentiated mouse EC, EG and ES cells
  • Expression of SSEA-3 is down regulated following differentiation of ES and EC cells
  • In contrast, the differentiation of mouse ES and EC cells may be accompanied by an increase in SSEA-3 expression
  • This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860) and has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog #18166)

Specifications

Antigen SSEA-3
Applications Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MC-631
Conjugate PE
Host Species Rat
Quantity 100 Tests
Primary or Secondary Primary
Target Species Rhesus Monkey, Human, Mouse, Rat
Isotype IgM κ
Cell Type Pluripotent Stem Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.