missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-3 Rat, anti-Mouse, Alexa Fluor 488, Clone: MC-631, STEMCELL Technologies
Rat Monoclonal Antibody
£283.50 - £633.15
Specifications
| Antigen | SSEA-3 |
|---|---|
| Clone | MC-631 |
| Applications | Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17168781
|
Stemcell Technologies
60061AD.1 |
25 Tests |
£283.50
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17158781
|
Stemcell Technologies
60061AD |
100 Tests |
£633.15
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The MC-631 antibody reacts with a galactosylgloboside epitope on the stage-specific embryonic antigen-3 (SSEA-3), which is expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey ES cell lines
- No immunoreactivity is evident with undifferentiated mouse EC, EG and ES cells
- Expression of SSEA-3 is down regulated following differentiation of ES and EC cells
- In contrast, the differentiation of mouse ES and EC cells may be accompanied by an increase in SSEA-3 expression
- This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860) and has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog #18166)
Specifications
| SSEA-3 | |
| Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| Rhesus Monkey, Human, Mouse, Rat | |
| Primary |
| MC-631 | |
| Monoclonal | |
| Rat | |
| IgM κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title