missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-3 Antibody (MC-631), Novus Biologicals™
Rat Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB100-1832
This item is not returnable.
View return policy
Description
SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| SSEA-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 10-20 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready | |
| Rat | |
| IgM purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| MC-631 | |
| Unconjugated | |
| Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3 | |
| 4-8 cell stage mouse embryos. | |
| 0.1 mL | |
| Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
| Recognizes a carbohydrate epitope of SSEA-3 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction