missing translation for 'onlineSavingsMsg'
Learn More

SSEA-1 Antibody (SPM490) - Azide and BSA Free, Novus Biologicals™

Product Code. 18380983 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380983 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380983 Supplier Novus Biologicals Supplier No. NBP311606

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone SPM490
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Gene Alias alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Host Species Mouse
Immunogen Myelomonocytic leukemia cells
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.