missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (CL6365), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £433.00
Specifications
| Antigen | SSEA-1 |
|---|---|
| Clone | CL6365 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626141
|
Novus Biologicals
NBP2-76491-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355861
|
Novus Biologicals
NBP2-76491 |
100 μL |
£433.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot.Specifications
| SSEA-1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| FUT4 | |
| IgG2b | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL6365 | |
| Monoclonal | |
| Purified | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 2526.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RKPSGAGWEKEWAEAPQEA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title