Learn More
SSEA-1 Antibody (CL6334) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76487MFV610
Product Code. 30299711
  • £591.00 / 0.10mL
Description
SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSEA-1
Monoclonal
mFluor Violet 610 SE
alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
This antibody was developed using a synthetic peptide derived from P22083, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
2526
Store at 4⪚C in the dark.
IgG2a
Western Blot
CL6334
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified