missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (CL6334) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76487APC
This item is not returnable.
View return policy
Description
SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human samples. It is validated for Western Blot
Specifications
| SSEA-1 | |
| Monoclonal | |
| APC | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| This antibody was developed using a synthetic peptide derived from P22083, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| 2526 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot | |
| CL6334 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction