missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSBP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15789710 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15789710 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15789710 Supplier Invitrogen™ Supplier No. PA551759

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LQVLRQFVRH ESETTTSLVL ERSLNRVHLL GRVGQDPVLR QVEGKNPVTI FSLATNEMWR SGDSEVYQLG DVSQKTTWHR ISVFRPGLRD VAYQYVKKGS RIYLEGKIDY GEYMDKNNVR RQATTIIADN IIFLSDQTKE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ssbp1
Gene Accession No. P28042, Q04837, Q9CYR0
Gene Alias 2810480P10Rik; G630031O20Rik; mtDBP; mtSSB; Mt-SSB; PWP1-interacting protein 17; single strand DNA-binding protein P16; single stranded DNA binding protein 1; single-stranded DNA binding protein 1; single-stranded DNA binding protein 1, mitochondrial; single-stranded DNA-binding protein, mitochondrial; SOSS-B1; Ssbp; Ssbp1
Gene Symbols Ssbp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSBP1. Recombinant protein control fragment (Product #RP-102227).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381760, 54304, 6742
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.