missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSBIP1/SOSS-C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81682
This item is not returnable.
View return policy
Description
SSBIP1/SOSS-C Polyclonal specifically detects SSBIP1/SOSS-C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SSBIP1/SOSS-C | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| INIP | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KEKRKLLMQNQSSTNHPGASIALSRPSLNKDFRDHAEQQHIAAQQKAALQHAHAHSSGYFITQDSAFGNL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.2mg/mL | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| chromosome 9 open reading frame 80, HSPC043, hSSB-interacting protein 1, hSSBIP1, minute INTS3/hSSB-associated element, MISE, Sensor of single-strand DNA complex subunit C, Sensor of ssDNA subunit C, Single-stranded DNA-binding protein-interacting protein 1, SOSS complex subunit C, SOSSC, SOSS-C, SSB-interacting protein 1, SSBIP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 58493 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction