Learn More
Invitrogen™ SSB Monoclonal Antibody (K70030_20C1), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | SSB |
|---|---|
| Clone | K70030_20C1 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30197048
|
Invitrogen™
MA555732 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211531
|
Invitrogen™
MA556112 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: MAENGDNEKM AALEAKICHQ IEYYFGDFNL PRDKFLKEQI KLDEGWVPLE IMIKFNRLNR LTTDFNVIVE ALSKSKAELM EISEDKTKIR RSPSKPLPEV TDEYKNDVKN RSVYIKGFPT DATLDDIKEW LEDKGQVLNI QMRRTLHKAF KGSIFVVFDS IESAKKFVET PGQKYKETDL LILFKDDYFA KKNEERKQNK VEAKLRAKQE QEAKQKLEED AEMKSLEEKI GCLLKFSGDL DDQTCREDLH ILFSNHGEIK WIDFVRGAKE GIILFKEKAK EALGKAKDAN NGNLQLRNKE VTWEVLEGEV EKEALKKIIE DQQESLNKWK SKGRRFKGKG KGNKAAQPGS GKGKVQFQGK KTKFASDDEH DEHDENGATG PVKRAREETD KEEPASKQQK TENGAGDQ
SSB Monoclonal antibody specifically detects SSB in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| SSB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein | |
| Recombinant human SSB (immunogen range is 1-408). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SSB |
| K70030_20C1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05455 | |
| 6741 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.