missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSB Antibody (OTI2C8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74356CL1
This item is not returnable.
View return policy
Description
SSB Monoclonal antibody specifically detects SSB in Canine,Human,Monkey,Primate samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SSB | |
| Monoclonal | |
| CoraFluor 1 | |
| La autoantigen, La ribonucleoprotein, lupus La protein, member 3, Sjoegren syndrome type B antigen, Sjogren syndrome antigen B (autoantigen La), SS-B, SS-B/La protein | |
| Full-length protein expressed in 293T cell transfected with human SSB expression vector | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6741 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2C8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction