missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20486
This item is not returnable.
View return policy
Description
SS18L1 Polyclonal specifically detects SS18L1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| SS18L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS pH7, 20% Glycerol with 0.025% Proclin 300 | |
| calcium-responsive transactivator, CRESTMGC78386, KIAA0693MGC26711, LP2261, SS18-like protein 1, synovial sarcoma translocation gene on chromosome 18-like 1, SYT homolog 1 | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Primary | |
| Species reactivity based on sequence identity: Cow (83%), Rat (81%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.73mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O75177 | |
| SS18L1 | |
| Recombinant protein encompassing a sequence within the center region of human CREST. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 26039 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction