missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SS18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563167
This item is not returnable.
View return policy
Description
Immunogen sequence: GQQPPMGMMG QVNQGNHMMG QRQIPPYRP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
The transcriptional coactivator SYT (synovial translocation protein) contains a conserved amino terminal SNH domain and a carboxy terminal QPGY domain, which is a functioning transcriptional activating sequence. Synovial sarcoma translocation (SSX) proteins, including SSX1-5, are transcriptional repressors that contain a repressor domain in their carboxy termini. SSX proteins are localized to the nucleus and expressed in testis and several types of cancers and, therefore, they are classified as C/T (cancer/testis) antigens. The t(x;18) translocation results in the fusion of the amino terminus of SYT to the carboxy terminus of either SSX1 or SSX2; both fusions result in the production of transcriptional activators.
Specifications
| SS18 | |
| Polyclonal | |
| Unconjugated | |
| SS18 | |
| D130059H17; MGC116875; Protein SSXT; Protein SYT; SS18; SS18, nBAF chromatin remodeling complex subunit; Ssxt; synovial sarcoma associated SS18-beta; synovial sarcoma associated SS18-delta; synovial sarcoma associated SS18-gamma; synovial sarcoma translocated to X chromosome protein; synovial sarcoma translocation; synovial sarcoma translocation, chromosome 18; synovial sarcoma, translocated to X chromosome; synovial sarcoma-associated Ss18-alpha; SYT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15532 | |
| SS18 | |
| Recombinant protein corresponding to Human SS18. Recombinant protein control fragment (Product #RP-101319). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction