missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRY Antibody (OTI3C8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74352
This item is not returnable.
View return policy
Description
SRY Monoclonal specifically detects SRY in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SRY | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| essential protein for sex determination in human males, sex determining region Y, sex-determining region on Y, sex-determining region Y protein, TDFsex determining region protein, TDY, Testis-determining factor | |
| Mouse | |
| 23.7 kDa | |
| 100 μg | |
| 6736 | |
| Human | |
| Lyophilized |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3C8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SRY | |
| Full length human recombinant protein of human SRY(NP_003131) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction