missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRXN1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-36991
This item is not returnable.
View return policy
Description
SRXN1 Polyclonal specifically detects SRXN1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SRXN1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C20orf139, chromosome 20 open reading frame 139, dJ850E9.2, EC 1.8.98.2, Npn3, SRX, SRX1, sulfiredoxin 1, sulfiredoxin 1 homolog, sulfiredoxin 1 homolog (S. cerevisiae), sulfiredoxin-1, YKL086W | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 140809 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_542763.1 | |
| SRXN1 | |
| Peptide with sequence C-KVQSLVDTIREDPD corresponding to internal region according to NP_542763.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction