missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRT 2104, MedChemExpress
Description
SRT 2104 is a first-in-class, highly selective and brain-permeable activator of the NAD+ dependent deacetylase Sirt1, increases Sirt1 protein, but shows no effect on Sirt1 mRNA. Used in the research of diabetes mellitus and Huntington’s disease.
Specifications
Specifications
| Chemical Name or Material | SRT 2104 |
| CAS | 1093403-33-8 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H24N6O2S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 5 mg/mL (9.68 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(C)N=C(C2=CC=CN=C2)S1)NC3=CC=CC=C3C4=CN5C(SC=C5CN6CCOCC6)=N4 |
| Molecular Weight (g/mol) | 516.64 |
| Formula Weight | 516.64 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?