missing translation for 'onlineSavingsMsg'
Learn More

SRT 2104, MedChemExpress

Product Code. 16431766
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431766

Brand: Medchem Express HY15262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SRT 2104 is a first-in-class, highly selective and brain-permeable activator of the NAD+ dependent deacetylase Sirt1, increases Sirt1 protein, but shows no effect on Sirt1 mRNA. Used in the research of diabetes mellitus and Huntington’s disease.

SRT 2104 is a first-in-class, highly selective and brain-permeable activator of the NAD+ dependent deacetylase Sirt1, increases Sirt1 protein, but shows no effect on Sirt1 mRNA. Used in the research of diabetes mellitus and Huntington’s disease.

Specifications

Chemical Name or Material SRT 2104
CAS 1093403-33-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H24N6O2S2
Quantity 500 mg
Solubility Information DMSO : 5 mg/mL (9.68 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N=C(C2=CC=CN=C2)S1)NC3=CC=CC=C3C4=CN5C(SC=C5CN6CCOCC6)=N4
Molecular Weight (g/mol) 516.64
Formula Weight 516.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.