missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRT 1720, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SRT 1720 is a selective activator of human SIRT1 with an EC1.5 of 0.16 μM, and shows less potent activities agaiinst SIRT2 and SIRT3 with EC1.5s of 37 μM and > 300 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | SRT 1720 |
| CAS | 925434-55-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H23N7OS |
| Quantity | 10 mg |
| Solubility Information | DMSO : 62.5 mg/mL (133.10 mM; ultrasonic and adjust pH to 5 with HCl) |
| SMILES | O=C(NC1=C(C=CC=C1)C2=CN3C(SC=C3CN4CCNCC4)=N2)C5=NC6=C(N=C5)C=CC=C6 |
| Molecular Weight (g/mol) | 469.56 |
| Formula Weight | 469.56 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction