missing translation for 'onlineSavingsMsg'
Learn More

SRT 1720 Hydrochloride, MedChemExpress

Product Code. 16482437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482437 50 mg 50mg
16488907 5 mg 5mg
16426646 100 mg 100mg
16426328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482437 Supplier Medchem Express Supplier No. HY15145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SRT 1720 Hydrochloride is a selective activator of SIRT1 with an EC50 of 0.10 μM, and shows less potent activities on SIRT2 and SIRT3.

SRT 1720 Hydrochloride is a selective activator of SIRT1 with an EC50 of 0.10 μM, and shows less potent activities on SIRT2 and SIRT3.

Specifications

Chemical Name or Material SRT 1720 Hydrochloride
CAS 2060259-60-9
Color Red
Purity Grade Notes Research
Molecular Formula C25H24ClN7OS
Quantity 50 mg
Solubility Information DMSO : 20 mg/mL (39.52 mM; Need ultrasonic) ∣H2O : 15.7 mg/mL (31.03 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC2=CC=CC=C2N=C1)NC3=C(C=CC=C3)C4=CN5C(CN6CCNCC6)=CSC5=N4.Cl
Molecular Weight (g/mol) 506.02
Formula Weight 506.02
Percent Purity 98.21%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.