missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRSF6 Recombinant Rabbit Monoclonal Antibody (24GB1825)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen SRSF6
Clone 24GB1825
Concentration 0.3 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275240
GREENER_CHOICE
View Documents
Invitrogen™
MA557265
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275390
View Documents
Invitrogen™
MA557266
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SRSF6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
Specifications

Specifications

SRSF6
0.3 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
G3V6S8, Q13247, Q3TWW8
362264, 6431, 67996
A synthesized peptide derived from human SRSF6 (aa 164-199).
Antibody
24GB1825
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Srsf6
1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6
Srsf6
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.