Learn More
Invitrogen™ SRSF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA589933
Product Code. 16567611
  • £441.00 / 100µL
Description
Immunogen sequence: MPRVYIGRLS YNVREKDIQR FFSGYGRLLE VDLKNGYGFV EFEDSRDADD AVYELNGKEL CGERVIVEHA RGPRRDRDGY SYGSRSGGGG YSSRRTSGRD; Positive Samples: HT-29, K562, HeLa, HepG2; Cellular Location: Nucleus, Nucleus speckle.
SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SRSF6
Polyclonal
Unconjugated
Srsf6
1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6
Rabbit
Affinity Chromatography
RUO
362264, 6431, 67996
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.93 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
G3V6S8, Q13247, Q3TWW8
Srsf6
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SRSF6 (NP_0062662).
100 μL
Primary
Human, Mouse, Rat