missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541810
This item is not returnable.
View return policy
Description
Peptide sequence: KERTNEGVIE FRSYSDMKRA LDKLDGTEIN GRNIRLIEDK PRTSHRRSYS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 82%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
Specifications
| SRSF6 | |
| Polyclonal | |
| Unconjugated | |
| Srsf6 | |
| 1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6431 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13247 | |
| Srsf6 | |
| Synthetic peptide directed towards the middle region of human SFRS6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction